Merck
  • Influence of the substituent on the phosphine ligand in novel rhenium(i) aldehydes. Synthesis, computational studies and first insights into the antiproliferative activity.
Influence of the substituent on the phosphine ligand in novel rhenium(i) aldehydes. Synthesis, computational studies and first insights into the antiproliferative activity.
Dalton transactions (Cambridge, England : 2003) (2018-09-19)
Michelle Muñoz-Osses, Daniel Siegmund, Alejandra Gómez, Fernando Godoy, Angélica Fierro, Leonel Llanos, Daniel Aravena, Nils Metzler-Nolte
ABSTRACT
Cyrhetrenyl aldehyde derivatives [(η5-C5H4CHO)Re(CO)2PR3] with R = methyl (Me, 2a), phenyl (Ph, 2b), and cyclohexyl (Cy, 2c) were synthesized by a photochemical reaction from the starting material [(η5-C5H4CHO)Re(CO)3] (1) and the corresponding phosphines. The complexes were fully characterized by FT-IR, 1H, 13C and 31P NMR spectroscopy, elemental analysis and mass spectrometry. The molecular structures of 2a-c have also been determined. Electronic structure calculations by TD-DFT and electrochemical studies are in sound agreement with the effect of the substitution of one carbonyl group by a phosphine ligand. Additionally, the antiproliferative activity of complexes 1 and 2a-c was studied on the human cancer cell lines HT-29 and PT-45 using an MTT assay. Out of all new compounds, only the triphenylphosphine derivative 2b exhibited pronounced cytotoxic effects on both cell lines, being ca. 1.5 times more potent than cisplatin.
Sigma-Aldrich
2-(Di-tert-butyl-phosphino)-1-phenyl-1H-pyrrole, 95%