Merck
  • The renal vasodilatory effect of prostaglandins is ameliorated in isolated-perfused kidneys of endotoxemic mice.
The renal vasodilatory effect of prostaglandins is ameliorated in isolated-perfused kidneys of endotoxemic mice.
Pflugers Archiv : European journal of physiology (2018-07-22)
Manuel Meurer, Katharina Ebert, Frank Schweda, Klaus Höcherl
ABSTRACT
Endotoxemia-related acute kidney injury (AKI) is associated with increased formation of prostaglandins, which may serve as a compensatory mechanism to maintain renal function. We hypothesized that an increase of renal EP2 or EP4 receptors and/or a downregulation of renal EP1 and EP3 receptors enhances PGE2-induced renal vasodilatation. Injection of lipopolysaccharide (LPS; 3 mg/kg i.p.) increased microsomal prostaglandin E synthase (mPGES)-1 and prostacyclin synthase expression, whereas mPGES-2 expression was unaltered. Further, LPS increased the mRNA abundance for the prostaglandin EP4 receptor, whereas the expressions of the EP1 and EP3 receptors were decreased. In isolated-perfused kidneys from control mice, PGE2 exerted a dual effect on renal vascular tone, inducing vasodilatation at lower concentrations and vasoconstriction at higher concentrations. In kidneys from endotoxemic mice, the vasodilatory component was more pronounced, whereas the vasoconstriction at higher PGE2 concentrations was absent. Similarly, prostacyclin (PGI2)-induced vasodilatation was more pronounced in endotoxemic kidneys. The enhanced vasodilatory effect was paralleled by an increase in renal vascular EP4 and prostacyclin IP receptor mRNA expression. Further, stimulation of renin secretion rate by PGE2 and PGI2 was enhanced in endotoxemic kidneys. Pretreatment with the cyclooxygenase (COX)-2 inhibitor SC-236 (10 mg/kg) did not alter the basal GFR, but augmented the LPS-induced decline in GFR, and attenuated the LPS-induced increase in plasma renin concentration in vivo. Our data suggest that an activation of the COX-2/mPGES-1 synthetic pathway is responsible for the increased renal formation of PGE2 in response to LPS and that the vasodilatory effect of PGE2 and PGI2 is enhanced during endotoxemia.
Sigma-Aldrich
SC-236, ≥98% (HPLC)