Merck
  • Influence of cycle stage, age and endometrial biopsy score on oxytocin receptor distribution and gene expression in the cervix and uterus of non-pregnant mares.
Influence of cycle stage, age and endometrial biopsy score on oxytocin receptor distribution and gene expression in the cervix and uterus of non-pregnant mares.
Theriogenology (2018-08-04)
A Annandale, R M Stroehle, M L Schulman, K P Sibeko-Matjila, G T Fosgate, J Handler, D C Vemming, S J Clift
ABSTRACT
Persistent breeding-induced endometritis (PBIE) or delayed uterine clearance (DUC) are major causes of mare subfertility. Oxytocin and its receptor are thought to play significant roles in the pathogenesis of DUC but the specific roles of oxytocin receptor (OR) distribution and gene expression remain undefined. In this study both OR distribution and gene expression in the endometrium, myometrium and cervix during both luteal and non-luteal phases in non-pregnant mares (n = 27) of differing age (young: 2-9 years, n = 17; old: > 10 years, n = 10) and endometrial biopsy score were described using immunohistochemistry (IHC) and quantitative reverse-transcription polymerase chain reaction (RT-qPCR), respectively. Immunohistochemistry showed a similar pattern of OR distribution in uterus and cervix, with the exception of the glandular epithelium, absent in the cervix. Uterine ORs were localized in endometrial luminal and glandular epithelia, transmural vascular endothelium, sub-epithelial and peri-glandular stromal cells and myometrial smooth muscle cells. The OR labeling intensity was consistently greatest in the vascular endothelium. Real-time qPCR showed a higher OR gene expression in myometrium compared to cervix (P = 0.001) and endometrium (P = 0.009). There was no difference in OR gene expression between cervix and endometrium (P = 1.0). Oxytocin receptor gene expression was significantly higher during the non-luteal phase in both combined uterine tissues (endometrium and myometrium) and myometrium. Oxytocin receptor distribution and gene expression were not influenced by a mare's age or endometrial biopsy score. As endometrial biopsy score and mare age were not predictors of OR gene expression, deficient OR gene expression is unlikely to be associated with DUC.
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Protease from Streptomyces griseus, Type XIV, ≥3.5 units/mg solid, powder