Merck
  • Synthesis of an oleic acid based pH-responsive lipid and its application in nanodelivery of vancomycin.
Synthesis of an oleic acid based pH-responsive lipid and its application in nanodelivery of vancomycin.
International journal of pharmaceutics (2018-08-21)
Danford Mhule, Rahul S Kalhapure, Mahantesh Jadhav, Calvin A Omolo, Sanjeev Rambharose, Chunderika Mocktar, Sanil Singh, Ayman Y Waddad, Valence M K Ndesendo, Thirumala Govender
ABSTRACT
Stimuli-responsive nano-drug delivery systems can optimize antibiotic delivery to infection sites. Identifying novel lipids for pH responsive delivery to acidic conditions of infection sites will enhance the performance of nano-drug delivery systems. The aim of the present investigation was to synthesize and characterize a biosafe novel pH-responsive lipid for vancomycin delivery to acidic conditions of infection sites. A pH-responsive solid lipid, N-(2-morpholinoethyl) oleamide (NMEO) was synthesized and used to prepare vancomycin (VCM)-loaded solid lipid nanoparticles (VCM_NMEO SLNs). The particle size (PS), polydispersity index (PDI), zeta potential (ZP) and entrapment efficiency (EE) of the formulation were 302.8 ± 0.12 nm, 0.23 ± 0.03, -6.27 ± 0.017 mV and 81.18 ± 0.57% respectively. The study revealed that drug release and antibacterial activity were significantly greater at pH 6.0 than at pH 7.4, while the in silico studies exposed the molecular mechanisms for improved stability and drug release. Moreover, the reduction of MRSA load was 4.14 times greater (p < 0.05) in the skin of VCM_NMEO SLNs treated mice than that of bare VCM treated specimens. Thus, this study confirmed that NMEO can successfully be used to formulate pH-responsive SLNs with potential to enhance the treatment of bacterial infections.
Sigma-Aldrich
4-(2-Aminoethyl)morpholine, 99%