Merck
  • Hemodynamic effects of endobronchial application of ornipressin versus terlipressin.
Hemodynamic effects of endobronchial application of ornipressin versus terlipressin.
Respiration; international review of thoracic diseases (2004-08-19)
Claudia Tüller, David Tüller, Michael Tamm, Martin H Brutsche
ABSTRACT
Ornipressin and terlipressin, two ADH-derivates, are instilled endobronchially for bronchoscopy- related bleeding without sufficient evidence of efficacy, nor safety. To compare the immediate hemodynamic effects of ornipressin and terlipressin used for procedure-related bleeding during flexible bronchoscopy. This retrospective study included patients referred for flexible bronchoscopy who needed administration of ornipressin (15 patients) or terlipressin (15 patients) for procedure-related bleeding. Endobronchial ornipressin (5 IU) or terlipressin (0.5 mg) was administered through the bronchoscope when bleeding persisted for 2 min of continuous suction, or when bleeding was considered increasing in time or major. Hemodynamic data were collected before, during and after administration of either drug. Biopsy-associated hemoptysis requiring medical treatment occurred in 110 (4.2%) of 2,592 bronchoscopies. After administration of ornipressin, no significant change in heart rate or blood pressure was observed. Following terlipressin instillation, heart rate increased from 93 +/- 17 to 101 +/- 22 b.p.m. (p = 0.03), and blood pressure decreased from 107 +/- 14 to 101 +/- 17 mm Hg (p = 0.04). Oxygen saturation under supplemental nasal oxygen was not different in both groups. None of the 110 patients died from bronchoscopy-related hemoptysis or needed further intervention to stop bleeding or prolonged monitoring because of hemodynamic instability. In contrast to ornipressin, administration of terlipressin is associated with significant changes in heart rate and blood pressure. However, these changes are of minor clinical importance, and terlipressin can be safely given for bronchoscopy-associated bleeding.
Sigma-Aldrich
Ornipressin acetate salt, ≥98% (HPLC)