Merck
  • Effects of S 38093, an antagonist/inverse agonist of histamine H3 receptors, in models of neuropathic pain in rats.
Effects of S 38093, an antagonist/inverse agonist of histamine H3 receptors, in models of neuropathic pain in rats.
European journal of pain (London, England) (2017-09-07)
T Chaumette, E Chapuy, E Berrocoso, M Llorca-Torralba, L Bravo, J A Mico, M Chalus, A Eschalier, D Ardid, F Marchand, A Sors
ABSTRACT
Histamine H3 receptors are mainly expressed on CNS neurons, particularly along the nociceptive pathways. The potential involvement of these receptors in pain processing has been suggested using H3 receptor inverse agonists. The antinociceptive effect of S 38093, a novel inverse agonist of H3 receptors, has been evaluated in several neuropathic pain models in rat and compared with those of gabapentin and pregabalin. While S 38093 did not change vocalization thresholds to paw pressure in healthy rats, it exhibited a significant antihyperalgesic effect in the Streptozocin-induced diabetic (STZ) neuropathy model after acute and chronic administration and, in the chronic constriction injury (CCI) model only after chronic administration, submitted to the paw-pressure test. Acute S 38093 administration at all doses tested displayed a significant cold antiallodynic effect in a model of acute or repeated administration of oxaliplatin-induced neuropathy submitted to cold tail immersion, cold allodynia being the main side effect of oxaliplatin in patients. The effect of S 38093 increased following chronic administration (i.e. twice a day during 5 days) in the CCI and STZ models except in the oxaliplatin models where its effect was already maximal from the first administration The kinetics and size of effect of S 38093 were similar to gabapentin and/or pregabalin. Finally, the antinociceptive effect of S 38093 could be partially mediated by α2 adrenoreceptors desensitization in the locus coeruleus. These results highlight the interest of S 38093 to relieve neuropathic pain and warrant clinical trials especially in chemotherapeutic agent-induced neuropathic pain. S 38093, a new H3 antagonist/inverse agonist, displays antiallodynic and antihyperalgesic effect in neuropathic pain, especially in oxaliplatin-induced neuropathy after chronic administration. This effect of S 38093 in neuropathic pain could be partly mediated by α2 receptors desensitization in the locus coeruleus.
Sigma-Aldrich
S 38093-2, ≥98% (HPLC)