Skip to Content
Merck
Xylazine hydrochloride European Pharmacopoeia (EP) Reference Standard

Xylazine hydrochloride

Xylazine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(2,6-Dimethylphenylamino)-5,6-dihydro-4H-thiazine hydrochloride, 5,6-Dihydro-2-(2,6-xylidino)-4H-1,3-thiazine hydrochloride, Xylazine hydrochloride
Empirical Formula (Hill Notation):
C12H16N2S · HCl
CAS Number:
Molecular Weight:
256.79

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade