![Trapidil impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/427/834/73e1de12-fe97-4719-a0de-978bf2e47657/640/73e1de12-fe97-4719-a0de-978bf2e47657.png)
Trapidil impurity A
![Trapidil impurity A European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/427/834/73e1de12-fe97-4719-a0de-978bf2e47657/640/73e1de12-fe97-4719-a0de-978bf2e47657.png)
Synonym(s):
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 5-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 6-Methyl-4-hydroxy-1,3,3a,7-tetraazaindene, 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, NSC 32071, NSC 511493
Empirical Formula (Hill Notation):
C6H6N4O
CAS Number:
Molecular Weight:
150.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing