![trans,trans-2,4-Decadienal technical grade, 85%](/deepweb/assets/sigmaaldrich/product/structures/576/790/4c820961-4766-437a-b4a4-9c9fa311abc4/640/4c820961-4766-437a-b4a4-9c9fa311abc4.png)
trans,trans-2,4-Decadienal
![trans,trans-2,4-Decadienal technical grade, 85%](/deepweb/assets/sigmaaldrich/product/structures/576/790/4c820961-4766-437a-b4a4-9c9fa311abc4/640/4c820961-4766-437a-b4a4-9c9fa311abc4.png)
Linear Formula:
CH3(CH2)4CH=CHCH=CHCHO
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing