Skip to Content
Merck
Thiodigalactoside ≥98% (HPLC)

Thiodigalactoside

Thiodigalactoside ≥98% (HPLC)
Synonym(s):
Thiodigalactoside, D-Galactopyranosyl-β-D-thiogalactopyranoside
Empirical Formula (Hill Notation):
C12H22O10S
CAS Number:
Molecular Weight:
358.36

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing