Skip to Content
Merck
Terpinolene technical, ≥85% (GC)

Terpinolene

Terpinolene technical, ≥85% (GC)
Synonym(s):
4-Isopropylidene-1-methylcyclohexene, p-Menth-1,4(8)-diene, p-Meth-1-en-8-yl-formate
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing