Skip to Content
Merck
Sterigmatocystin

Sterigmatocystin

Sterigmatocystin
Synonym(s):
3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furol[3′,2′:4,5]furo[2,3-c]xanthen-7-one
Empirical Formula (Hill Notation):
C18H12O6
CAS Number:
Molecular Weight:
324.28
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing