![S-Selenomethionine certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/322/737/df924801-8337-442e-ac4f-dcedc039384d/640/df924801-8337-442e-ac4f-dcedc039384d.png)
S-Selenomethionine
![S-Selenomethionine certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/structures/322/737/df924801-8337-442e-ac4f-dcedc039384d/640/df924801-8337-442e-ac4f-dcedc039384d.png)
Synonym(s):
Seleno-L-methionine, (S)-2-Amino-4-(methylseleno)butyric acid
Linear Formula:
CH3SeCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
196.11
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing