![(S)-(−)-N-Benzyl-α-methylbenzylamine 99%](/deepweb/assets/sigmaaldrich/product/structures/364/186/f071e633-8719-43f1-8931-ae65c92c13f5/640/f071e633-8719-43f1-8931-ae65c92c13f5.png)
(S)-(−)-N-Benzyl-α-methylbenzylamine
![(S)-(−)-N-Benzyl-α-methylbenzylamine 99%](/deepweb/assets/sigmaaldrich/product/structures/364/186/f071e633-8719-43f1-8931-ae65c92c13f5/640/f071e633-8719-43f1-8931-ae65c92c13f5.png)
Synonym(s):
(S)-(−)-N-(1-Phenylethyl)benzylamine, (S)-(−)-N-Benzyl-α-phenylethylamine
Linear Formula:
C6H5CH(CH3)NHCH2C6H5
CAS Number:
Molecular Weight:
211.30
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing