
Sildenafil Related Compound A

Synonym(s):
5-[2-Ethoxy-5[(4-methylpiperazin-1-yl)sulfonyl]phenyl]-1-methyl-3-(2-methylpropyl)-6,7-dihydro-1H-pyrazol[4,3-d]pyrimidin-7-one, Isobutyl sildenafil
Empirical Formula (Hill Notation):
C23H32N6O4S
CAS Number:
Molecular Weight:
488.60
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2