![Senegenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/386/614/7e8cb39f-68ed-40bc-888b-df6e1b93758f/640/7e8cb39f-68ed-40bc-888b-df6e1b93758f.png)
Senegenin
![Senegenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/386/614/7e8cb39f-68ed-40bc-888b-df6e1b93758f/640/7e8cb39f-68ed-40bc-888b-df6e1b93758f.png)
Synonym(s):
12-(Chloromethyl)-2,3-dihydroxy-27-norolean-13-ene-23,28-dioic acid, Tenuigenin
Empirical Formula (Hill Notation):
C30H45ClO6
CAS Number:
Molecular Weight:
537.13
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing