Sauchinone
Synonym(s):
(5aα,7α,8β,8aβ,14aS*,14bβ)-5a,6,7,8,8a,14b-Hexahydro-7,8-dimethyl-5H-benzo[kl]bis[1,3]dioxolo[4,5-b:4′,5′-g]xanthen-5-one, (5aR,7R,8S,8aR,14aS,14bR)-5a,6,7,8,8a,14b-Hexahydro-7,8-dimethyl-5H-Benzo[kl]bis[1,3]dioxolo[4,5-b:4′,5′-g]xanthen-5-one
Empirical Formula (Hill Notation):
C20H20O6
CAS Number:
Molecular Weight:
356.37
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing