Salicylaldehyde
Synonym(s):
2-Hydroxybenzaldehyde
Linear Formula:
2-(HO)C6H4CHO
CAS Number:
Molecular Weight:
122.12
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing