Skip to Content
Merck
Rizatriptan benzoate salt ≥98% (HPLC)

Rizatriptan benzoate salt

Rizatriptan benzoate salt ≥98% (HPLC)
Synonym(s):
MK-462, N,N-Dimethyl-2-[5-(1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]ethylamine benzoate salt
Empirical Formula (Hill Notation):
C15H19N5 · C7H6O2
CAS Number:
Molecular Weight:
391.47

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing