Skip to Content
Merck
vibrant-m

Protopanaxatriol

vibrant-m
Synonym(s):
20S-Protopanaxatriol, 20(S)-APPT, g-PPT
Empirical Formula (Hill Notation):
C30H52O4
CAS Number:
Molecular Weight:
476.73

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing