
Prochlorperazine Related Compound A

Synonym(s):
4-Chloro-10-[3-(4-methylpiperazin-1-yl)propyl]-10H-phenothiazine dihydrochloride
Empirical Formula (Hill Notation):
C20H24ClN3S · 2HCl
CAS Number:
Molecular Weight:
446.86
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2