Piroxicam Related Compound A
Synonym(s):
2-Aminopyridine, 2-AP, 2-Pyridinamine, 2-Pyridylamine
Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing