Skip to Content
Merck
Penicillin G Potassium Pharmaceutical Secondary Standard; Certified Reference Material

Penicillin G potassium

Penicillin G Potassium Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Penicillin G potassium salt, Benzylpenicillin potassium salt
Empirical Formula (Hill Notation):
C16H17KN2O4S
CAS Number:
Molecular Weight:
372.48

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade