![Pellitorine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/339/596/48b56c24-0f79-4d70-8501-ceaca6880fae/640/48b56c24-0f79-4d70-8501-ceaca6880fae.png)
Pellitorine
![Pellitorine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/339/596/48b56c24-0f79-4d70-8501-ceaca6880fae/640/48b56c24-0f79-4d70-8501-ceaca6880fae.png)
Synonym(s):
(2E,4E)-N-(2-Methylpropyl)-2,4-decadienamide, (2E,4E)-N-Isobutyl-2,4-decadienamide, 2E,4E-Decadienoic acid N-isobutylamide, trans-Pellitorin, trans-Pellitorine
Empirical Formula (Hill Notation):
C14H25NO
CAS Number:
Molecular Weight:
223.35
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing