
Paroxetine impurity D

Synonym(s):
(3R-trans)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine
Empirical Formula (Hill Notation):
C19H20FNO3 · HCl
CAS Number:
Molecular Weight:
365.83
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing