
Open Ring Aztreonam

Synonym(s):
(2S,3S)-2-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-3-(sulfoamino)butanoic acid, (2S,3S)-2-{(Z)-2-[2-Aminothiazol-4-yl]-2-[2-carboxypropan-2-yloxyimino]acetamido}-3-(sulfoamino)butanoic acid
Empirical Formula (Hill Notation):
C13H19N5O9S2
CAS Number:
Molecular Weight:
453.45
Products
Brand
Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2