Skip to Content
Merck
vibrant-m

Nicotine Related Compound A

vibrant-m
Synonym(s):
Anatabine, (RS)-Anatabine, (±)-Anatabine, 1,2,3,6-Tetrahydro-2,3′-bipyridine, 1,2,3,6-Tetrahydro-2,3′-dipyridine
Empirical Formula (Hill Notation):
C10H12N2
CAS Number:
Molecular Weight:
160.22

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1