![Nafcillin sodium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/273/055/138e8b2a-2247-42a8-a34e-806ca40e7ec9/640/138e8b2a-2247-42a8-a34e-806ca40e7ec9.png)
Nafcillin sodium
![Nafcillin sodium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/273/055/138e8b2a-2247-42a8-a34e-806ca40e7ec9/640/138e8b2a-2247-42a8-a34e-806ca40e7ec9.png)
Synonym(s):
Nafcillin sodium salt monohydrate, 6-(2-Ethoxy-1-naphthamido)penicillin
Linear Formula:
C21H21N2O5SNa · H2O
CAS Number:
Molecular Weight:
454.47
Products
Product Category
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing