Skip to Content
Merck
Myristoleic acid analytical standard

Myristoleic acid

Myristoleic acid analytical standard
Synonym(s):
cis-9-Tetradecenoic acid
Linear Formula:
CH3(CH2)3CH=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
226.36
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing