![Monoclonal Anti-FOXP2 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL5310, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/105/109/ba7ee20d-a7e0-4d06-9baf-3883a826b167/640/ba7ee20d-a7e0-4d06-9baf-3883a826b167.jpg)
Monoclonal Anti-FOXP2 antibody produced in mouse
![Monoclonal Anti-FOXP2 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL5310, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/105/109/ba7ee20d-a7e0-4d06-9baf-3883a826b167/640/ba7ee20d-a7e0-4d06-9baf-3883a826b167.jpg)
Synonym(s):
CAGH44, SPCH1, TNRC10
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing