![Monoclonal Anti-Biotin antibody produced in mouse clone BN-34, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/503/773/95b35bf5-1362-47cc-ac47-a4ddaf8c8807/640/95b35bf5-1362-47cc-ac47-a4ddaf8c8807.jpg)
Monoclonal Anti-Biotin antibody produced in mouse
![Monoclonal Anti-Biotin antibody produced in mouse clone BN-34, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/503/773/95b35bf5-1362-47cc-ac47-a4ddaf8c8807/640/95b35bf5-1362-47cc-ac47-a4ddaf8c8807.jpg)
Synonym(s):
Monoclonal Anti-Biotin
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Conjugate
Quality Segment
Application
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing