Skip to Content
Merck
Mastoparan, Vespula lewisii ≥97% (HPLC), powder

Mastoparan, Vespula lewisii

Mastoparan, Vespula lewisii ≥97% (HPLC), powder
Empirical Formula (Hill Notation):
C70H131N19O15
CAS Number:
Molecular Weight:
1478.91

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing