![Maneb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/409/944/c15e9356-c0db-47a4-a1e2-4760c08b6a6b/640/c15e9356-c0db-47a4-a1e2-4760c08b6a6b.png)
Maneb
![Maneb PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/409/944/c15e9356-c0db-47a4-a1e2-4760c08b6a6b/640/c15e9356-c0db-47a4-a1e2-4760c08b6a6b.png)
Synonym(s):
Manganese ethylenebis(dithiocarbamate)
Linear Formula:
[C4H6MnN2S4]n
CAS Number:
EC Number:
Products
Product Category
Brand
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing