![Magnolin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/368/607/e977b056-36cf-471d-9266-197daf1eb492/640/e977b056-36cf-471d-9266-197daf1eb492.png)
Magnolin
![Magnolin European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/368/607/e977b056-36cf-471d-9266-197daf1eb492/640/e977b056-36cf-471d-9266-197daf1eb492.png)
Synonym(s):
Magnolin, (1S,3aR,4S,6aR)-1-(3,4-Dimethoxyphenyl)tetrahydro-4-(3,4,5-trimethoxyphenyl)-1H,3H-furo[3,4-c]furan, Medioresinol dimethyl ether
Empirical Formula (Hill Notation):
C23H28O7
CAS Number:
Molecular Weight:
416.46
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing