Skip to Content
Merck
Lutetium(III) trifluoromethanesulfonate 98%

Lutetium(III) trifluoromethanesulfonate

Lutetium(III) trifluoromethanesulfonate 98%
Synonym(s):
Lutetium(III) triflate, Trifluoromethanesulfonic acid lutetium(III) salt
Linear Formula:
(CF3SO3)3Lu
CAS Number:
Molecular Weight:
622.17

Products

Brand
Formula Weight
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing