Skip to Content
Merck
L-Tryptophan-13C11,15N2 ≥99 atom %, ≥98% (CP)

L-Tryptophan-13C11,15N2

L-Tryptophan-13C11,15N2 ≥99 atom %, ≥98% (CP)
Synonym(s):
L-Tryptophan-ul-13C,15N
Empirical Formula (Hill Notation):
13C11H1215N2O2
CAS Number:
Molecular Weight:
217.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing