Liriope muscari Saponin C
Empirical Formula (Hill Notation):
C44H70O17
CAS Number:
Molecular Weight:
871.02
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing