Skip to Content
Merck
Leupeptin >4000 U/mg

Leupeptin

Leupeptin >4000 U/mg
Synonym(s):
Leupeptin hemisulfate salt, Acetyl-Leu-Leu-Arg-al, N-Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate salt
Linear Formula:
C20H38N6O4 · 1/2H2SO4
CAS Number:
Molecular Weight:
475.59

Products

Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Optical Isomer
Showing 1-5 of 5
Compare
Product Number
Description
Pricing