
Latanoprost Related Compound D

Synonym(s):
Isopropyl (Z)-7-{(1R,2R,3R,5S)-2-[(3R)-5-phenyl-3-(triethylsilyloxy)pentyl]-3,5-bis(triethylsilyloxy)cyclopentyl}hept-5-enoate, Latanoprost tris(triethylsilyl) ether
Empirical Formula (Hill Notation):
C44H82O5Si3
CAS Number:
Molecular Weight:
775.38
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing