Skip to Content
Merck
vibrant-m

Ketoprofen Related Compound D hydrochloride

vibrant-m
Synonym(s):
1-(3-Benzoylphenyl)ethanone, 3-Acetylbenzophenone
Empirical Formula (Hill Notation):
C15H12O2
CAS Number:
Molecular Weight:
224.25

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1