Isosilybin (A + B mixture)
Synonym(s):
Isosilybin, mixture of isomers, Isosilybinin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing