Skip to Content
Merck
Isobutylamine 99%

Isobutylamine

Isobutylamine 99%
Synonym(s):
1-Amino-2-methylpropane, Isobutylamine
Linear Formula:
(CH3)2CHCH2NH2
CAS Number:
Molecular Weight:
73.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing