![Iopamidol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/187/873/eb63df1b-9a82-46a3-8472-0052bb2f7e53/640/eb63df1b-9a82-46a3-8472-0052bb2f7e53.jpg)
Iopamidol
![Iopamidol Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/187/873/eb63df1b-9a82-46a3-8472-0052bb2f7e53/640/eb63df1b-9a82-46a3-8472-0052bb2f7e53.jpg)
Synonym(s):
(S)-N,N′-Bis[2-hydroxy-1-(hydroxymethyl)ethyl]-2,4,6-triiodo-5-lactamidoisophthalamide, Iopamidol
Empirical Formula (Hill Notation):
C17H22I3N3O8
CAS Number:
Molecular Weight:
777.09
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-3 of 3