Iodoquinol
Synonym(s):
5,7-Diiodo-8-hydroxyquinoline, 5,7-Diiodo-8-quinolinol
Empirical Formula (Hill Notation):
C9H5I2NO
CAS Number:
Molecular Weight:
396.95
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing