Skip to Content
Merck
Iodobenzene 98%

Iodobenzene

Iodobenzene 98%
Synonym(s):
Iodobenzene
Empirical Formula (Hill Notation):
C6H5I
CAS Number:
Molecular Weight:
204.01

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing