![I3,II8-Biapigenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/157/727/16140d1c-bbcd-49ef-a41a-e445c0d91346/640/16140d1c-bbcd-49ef-a41a-e445c0d91346.png)
I3,II8-Biapigenin
![I3,II8-Biapigenin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/157/727/16140d1c-bbcd-49ef-a41a-e445c0d91346/640/16140d1c-bbcd-49ef-a41a-e445c0d91346.png)
Synonym(s):
3,8″-Biapigenin, 5,5′,7,7′-Tetrahydroxy-2,2′-bis(4-hydroxyphenyl)-[3,8′-bi-4H-1-benzopyran]-4,4′-dione
Empirical Formula (Hill Notation):
C30H18O10
CAS Number:
Molecular Weight:
538.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing