![Hycon® Agar Strips LAC-y Tryptic Soy Agar, Beta-lactamase, Lecithin, Polysorbate 80, irradiated, double packed of 40 strips, suitable for air monitoring](/deepweb/assets/sigmaaldrich/product/images/108/074/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d/640/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d.jpg)
Hycon® Agar Strips LAC-y
![Hycon® Agar Strips LAC-y Tryptic Soy Agar, Beta-lactamase, Lecithin, Polysorbate 80, irradiated, double packed of 40 strips, suitable for air monitoring](/deepweb/assets/sigmaaldrich/product/images/108/074/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d/640/71e86c8b-36ce-4de1-9a4a-ad4917d97f8d.jpg)
Synonym(s):
Hycon Ready-to-use Agar Strips
Products
Product Category
Brand
Color
Physical Form
Agency
Chemical Composition
Compatibility
Quality Segment
Application
Microorganism Suitability
Package Size
Sterilization Method
Showing 1-1 of 1