Skip to Content
Merck
Higenamine hydrochloride phyproof® Reference Substance

Higenamine hydrochloride

Higenamine hydrochloride phyproof® Reference Substance
Synonym(s):
Higenamine hydrochloride, (±)-Demethylcoclaurine hydrochloride, (±)-Norcoclaurine hydrochloride
Empirical Formula (Hill Notation):
C16H17NO3 · HCl
CAS Number:
Molecular Weight:
307.77

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing