![GW9662 >98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/675/320/4bb04ed5-eb3b-4f3d-b10d-daafa9ddb4af/640/4bb04ed5-eb3b-4f3d-b10d-daafa9ddb4af.png)
GW9662
![GW9662 >98% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/675/320/4bb04ed5-eb3b-4f3d-b10d-daafa9ddb4af/640/4bb04ed5-eb3b-4f3d-b10d-daafa9ddb4af.png)
Synonym(s):
2-Chloro-5-nitro-N-phenylbenzamide
Empirical Formula (Hill Notation):
C13H9ClN2O3
CAS Number:
Molecular Weight:
276.68
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing