Skip to Content
Merck
Forsythiaside A ≥98% (HPLC)

Forsythiaside A

Forsythiaside A ≥98% (HPLC)
Synonym(s):
Forsythoside A, Forsythiaside
Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing